Assay Details
| Gene Symbol: Olfr218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001809.2 | NP_001001809.2 | 1 | 566 | 121 | |
| GenBank mRNA | BC104284.1 | - | 1 | 565 | 121 | |
| BC104291.1 | - | 1 | 566 | 40042773 | 121 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144150 | NM_001001809.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144150 | BC104284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144150 | BC104291.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159587 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144150 | NM_001001809.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144150 | BC104284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144150 | BC104291.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P303784 | NM_001001809.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P303784 | BC104284.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P303784 | BC104291.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836664 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159587 | - | - |
| Whole Mouse Genome G4122A | A_51_P303784 | NM_001001809.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303784 | BC104284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303784 | BC104291.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130935 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159587 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00803756 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00803750 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00803755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00399929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00803747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00859469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00399936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00859471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00751707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836656 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351862 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351861 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100013553.mm.1 | NM_001001809.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013553.mm.1 | NM_001001809.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013553.mm.1 | TC0100001628.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 554593 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161008 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161008 | - | - |
| mouse-6 v1.1 (Illumina) | GI_49227319-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161008 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161008 | - | - |

Olfr218
Genomic Map