Assay Details
Gene Symbol: B630019K06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045448.1 | 1 | 2387 | 125 | ||
GenBank mRNA | AK046767.1 | - | 1 | 2359 | 125 | |
BC055780.1 | - | 1 | 2280 | 6403722 | 125 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244418 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244415 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244419 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244417 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244416 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513062 | BC055780.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513062 | AK046767.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513062 | NM_175327.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513062 | BC055780.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513062 | AK046767.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513062 | NM_175327.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383518 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952113 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20545 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112385 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513062 | BC055780.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513062 | AK046767.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513062 | NM_175327.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513062 | BC055780.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513062 | AK046767.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513062 | NM_175327.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244410 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244421 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244420 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244413 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244423 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244411 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32244431 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112385 | - | - |
Whole Mouse Genome G4122A | A_51_P513062 | BC055780.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513062 | AK046767.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513062 | NM_175327.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513062 | BC055780.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513062 | AK046767.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513062 | NM_175327.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134661 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112385 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15125134 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14606355 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244415 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244417 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244418 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244416 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32244419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32244433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14538005 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14882182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14707810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244452 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15031304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14707799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14707807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244447 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244415 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244468 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244467 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244441 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244446 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14538006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15125134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244466 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244428 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244457 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244443 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15031307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32244414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14538011 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10598563 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598564 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1447618_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1433452_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10598562 | - | - |
Mouse Genome 430 2.0 Array | 1433452_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1447618_at | - | - |
Mouse Genome 430 2.0 Array | 1433452_at | - | - |
Mouse Genome 430 2.0 Array | 1447618_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102941_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102941_at | - | - |
Mouse Expression Set 430 | 1433452_PM_at | - | - |
Mouse Expression Set 430 | 1447618_at | - | - |
Mouse Expression Set 430 | 1447618_PM_at | - | - |
Mouse Expression Set 430 | 1433452_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447618_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1433452_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447618_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1433452_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 831793 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3850195 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003850195 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222840 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1222840 | - | - |
mouse-6 v1.1 (Illumina) | scl55053.1.80_33-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3850195 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl55053.1.80_33-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850195 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1222840 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222840 | - | - |
