Assay Details
Gene Symbol: Atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025381.2 | NP_079657.1 | 1 | 68 | 89 | |
GenBank mRNA | AK003419.1 | - | 1 | 65 | 89 | |
AK007327.1 | - | 1 | 69 | 89 | ||
AK152153.1 | - | 1 | 65 | 89 | ||
AK152929.1 | - | 1 | 92 | 89 | ||
AK152959.1 | - | 1 | 65 | 89 | ||
BC016553.1 | - | 1 | 78 | 2812592 | 89 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24326761 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24326760 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24326758 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973486 | BC016553.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973486 | AK152929.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973486 | AK152959.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973486 | NM_025381.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973486 | AK003419.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605647 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479174 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178386 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176747 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119522 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973486 | BC016553.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973486 | AK152929.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973486 | AK152959.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973486 | NM_025381.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973486 | AK003419.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P514094 | BC016553.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P514094 | AK152929.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P514094 | AK152959.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P514094 | NM_025381.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P514094 | AK003419.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310539 | BC016553.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310539 | AK152929.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310539 | AK152959.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310539 | NM_025381.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310539 | AK003419.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326710 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326713 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326686 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326683 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326696 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326695 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326687 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326705 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326694 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326684 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326681 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326701 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326717 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326678 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326711 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326704 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326691 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326680 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326715 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326698 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326722 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326692 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326716 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326719 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326677 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326707 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326720 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24326724 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178386 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176747 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119522 | - | - |
Whole Mouse Genome G4122A | A_52_P514094 | BC016553.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P514094 | AK152929.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P514094 | AK152959.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P514094 | NM_025381.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P514094 | AK003419.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P310539 | BC016553.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P310539 | AK152929.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P310539 | AK152959.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P310539 | NM_025381.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P310539 | AK003419.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124009 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128573 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176747 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119522 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178386 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05244765 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05244764 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05244762 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05635154 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24326761 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24326758 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24326760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24326765 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326722 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326678 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05509413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05443943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326763 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05759741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05332203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05563268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05642506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05637770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05244766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05423731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326715 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05759745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05509406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326673 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05466075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326758 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326762 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05445041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24326680 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10536896 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10452875 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10536897 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423993_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10536895 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10452874 | - | - |
Mouse Genome 430 2.0 Array | 1423993_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1423993_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66144_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66144_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600001592.mm.1 | TC0600000233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600001592.mm.1 | NM_025381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600001592.mm.1 | NM_025381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600001595.mm.1 | TC0600000233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600001595.mm.1 | NM_025381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600001595.mm.1 | NM_025381.2 | Refseq |
Mouse Expression Set 430 | 1423993_at | - | - |
Mouse Expression Set 430 | 1423993_PM_at | - | - |
Murine Genome U74v2 Set | 168588_at | - | - |
Murine Genome U74v2 Set | 170348_i_at | - | - |
Murine Genome U74v2 Set | 168588_PM_at | - | - |
Murine Genome U74v2 Set | 170348_PM_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1423993_at | - | - |
Mouse Genome 430A 2.0 Array | 1423993_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423993_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423993_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5420082 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005420082 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218325 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1218325 | - | - |
mouse-6 v1.1 (Illumina) | scl066144.2_329-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5420082 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066144.2_329-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420082 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1218325 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218325 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066144.2_329-S | - | - |
