Assay Details
| Gene Symbol: Olfr860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146528.2 | NP_666739.1 | 2 | 864 | 148 | |
| GenBank mRNA | BC120862.1 | - | 1 | 695 | 40129785 | 148 |
| BC137831.1 | - | 1 | 695 | 8860850 | 148 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P87763 | BC137831.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P87763 | NM_146528.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P87763 | BC120862.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P87763 | BC137831.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P87763 | NM_146528.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P87763 | BC120862.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138349 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P87763 | BC137831.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P87763 | NM_146528.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P87763 | BC120862.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P87763 | BC137831.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P87763 | NM_146528.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P87763 | BC120862.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P87763 | BC137831.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P87763 | NM_146528.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P87763 | BC120862.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P87763 | BC137831.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P87763 | NM_146528.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P87763 | BC120862.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26341068 | - | - |
| Whole Mouse Genome G4122A | A_52_P87763 | BC137831.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P87763 | NM_146528.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P87763 | BC120862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P87763 | BC137831.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P87763 | NM_146528.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P87763 | BC120862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499317 | NM_011001.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116522 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07470431 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07620565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07892772 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07620568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26341060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07703977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07703968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26341032 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591204 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 424887 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 770193 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770193 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2649548 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2649548 | - | - |
| mouse-6 v1.1 (Illumina) | scl6689.1.1_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770193 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl6689.1.1_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770193 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2649548 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2649548 | - | - |

Olfr860
Genomic Map