| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10486862 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486873 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486863 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486870 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486869 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486872 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486865 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486864 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486871 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486874 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486866 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486860 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486868 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10486861 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10486858 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10486867 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0200038114.mm.1 |
NM_001081975.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038114.mm.1 |
NM_001081975.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038114.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038123.mm.1 |
NM_026220.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038123.mm.1 |
NM_026220.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038123.mm.1 |
TC0200004492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038109.mm.1 |
NM_001081975.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038109.mm.1 |
NM_001081975.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038109.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038110.mm.1 |
NM_001081975.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038110.mm.1 |
NM_001081975.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038110.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038121.mm.1 |
NM_026220.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038121.mm.1 |
NM_026220.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038121.mm.1 |
TC0200004492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038106.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038112.mm.1 |
NM_001081975.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038112.mm.1 |
NM_001081975.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038112.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038113.mm.1 |
NM_001081975.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038113.mm.1 |
NM_001081975.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038113.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038111.mm.1 |
NM_001081975.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038111.mm.1 |
NM_001081975.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038111.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038124.mm.1 |
NM_026220.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038124.mm.1 |
NM_026220.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038124.mm.1 |
TC0200004492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038119.mm.1 |
NM_026220.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038119.mm.1 |
NM_026220.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038119.mm.1 |
TC0200004492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038107.mm.1 |
NM_001081975.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038107.mm.1 |
NM_001081975.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038107.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038108.mm.1 |
NM_001081975.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038108.mm.1 |
NM_001081975.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038108.mm.1 |
TC0200004491.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038118.mm.1 |
NM_026220.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038118.mm.1 |
NM_026220.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038118.mm.1 |
TC0200004492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038120.mm.1 |
NM_026220.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038120.mm.1 |
NM_026220.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038120.mm.1 |
TC0200004492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038122.mm.1 |
NM_026220.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038122.mm.1 |
NM_026220.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038122.mm.1 |
TC0200004492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200038125.mm.1 |
NM_026220.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038125.mm.1 |
NM_026220.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200038125.mm.1 |
TC0200004492.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1449444_PM_a_at |
- |
- |
| Mouse Expression Set 430 |
1449444_a_at |
- |
- |
| Mouse Expression Set 430 |
1449445_PM_x_at |
- |
- |
| Mouse Expression Set 430 |
1419370_a_at |
- |
- |
| Mouse Expression Set 430 |
1419370_PM_a_at |
- |
- |
| Mouse Expression Set 430 |
1449445_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1419370_PM_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1449444_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1449445_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1449444_PM_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1419370_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1449445_PM_x_at |
- |
- |