Assay Details
| Gene Symbol: Rpl27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011975.3 | NP_036105.2 | 5 | 1029 | 105 | |
| GenBank mRNA | AK010286.1 | - | 5 | 1030 | 105 | |
| BB734594.1 | - | 1 | 323 | 105 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408488 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408486 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408483 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408485 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408484 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065327 | X05021.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065327 | AK150907.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065327 | AK011361.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065327 | AK167792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065327 | AK167915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048671 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P132459 | BB734594.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P132459 | NM_011975.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P132459 | AA253983.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P132459 | AK010286.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P132459 | BB734594.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P132459 | NM_011975.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P132459 | AA253983.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P132459 | AK010286.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36906 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148175 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048671 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P132459 | BB734594.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P132459 | NM_011975.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P132459 | AA253983.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P132459 | AK010286.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P132459 | BB734594.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P132459 | NM_011975.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P132459 | AA253983.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P132459 | AK010286.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065327 | X05021.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065327 | AK150907.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065327 | AK011361.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065327 | AK167792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065327 | AK167915.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132459 | BB734594.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132459 | NM_011975.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132459 | AA253983.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132459 | AK010286.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132459 | BB734594.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132459 | NM_011975.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132459 | AA253983.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132459 | AK010286.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25408505 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148175 | - | - |
| Whole Mouse Genome G4122A | A_51_P418662 | NR_028079.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P418662 | AF357390.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P418662 | AK138110.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132459 | BB734594.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132459 | NM_011975.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132459 | AA253983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132459 | AK010286.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132459 | BB734594.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132459 | NM_011975.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132459 | AA253983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132459 | AK010286.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122990 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116442 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148175 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06425553 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06425545 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06425561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06425567 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06425563 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06425554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06425557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408483 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408488 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408485 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408484 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25408486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25408502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06425561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06179645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06425545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06286575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06257863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06159872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25408485 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10363480 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406855 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10578065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10474670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556203 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426661_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10363479 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10406854 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10556200 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547151 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10578064 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10487643 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10491062 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10357369 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10474669 | - | - |
| Mouse Genome 430 2.0 Array | 1426661_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426661_at | - | - |
| Mouse Genome 430 2.0 Array | 1419799_at | - | - |
| Mouse Genome 430 2.0 Array | 1419799_PM_at | - | - |
| Mouse Expression Set 430 | 1426661_PM_at | - | - |
| Mouse Expression Set 430 | 1419799_at | - | - |
| Mouse Expression Set 430 | 1419799_PM_at | - | - |
| Mouse Expression Set 430 | 1426661_at | - | - |
| Murine Genome U74v2 Set | 101680_PM_at | - | - |
| Murine Genome U74v2 Set | 97691_PM_at | - | - |
| Murine Genome U74v2 Set | 101573_f_at | - | - |
| Murine Genome U74v2 Set | 101573_PM_f_at | - | - |
| Murine Genome U74v2 Set | 101680_at | - | - |
| Murine Genome U74v2 Set | 97691_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426661_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426661_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419799_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419799_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 669461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6840494 | - | - |
| MouseWG-6 v2 Expression BeadChip | 110195 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 110195 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3130451 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6840494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006840494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130451 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215659 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221403 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221403 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215659 | - | - |
| mouse-6 v1.1 (Illumina) | scl32205.4_514-S | - | - |
| mouse-6 v1.1 (Illumina) | scl026451.5_30-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0026451.1_182-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6840494 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110195 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130451 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0026451.1_182-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl026451.5_30-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32205.4_514-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130451 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840494 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2882059 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221403 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221403 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2882059 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641304 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32205.4_514-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl026451.5_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0026451.1_182-S | - | - |

Rpl27a
Genomic Map