Assay Details
| Gene Symbol: Sumo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133354.2 | NP_579932.1 | 4 | 378 | 150 | |
| GenBank mRNA | AK012619.1 | - | 4 | 379 | 150 | |
| AK085238.1 | - | 4 | 325 | 150 | ||
| AK132213.1 | - | 377 | 150 | |||
| AK160404.1 | - | 4 | 395 | 150 | ||
| AK167308.1 | - | 4 | 388 | 150 | ||
| AK219519.1 | - | 2 | 86 | 150 | ||
| BC017522.1 | - | 4 | 352 | 4512364 | 150 | |
| BC083326.1 | - | 4 | 290 | 6819700 | 150 | |
| BC095930.1 | - | 4 | 365 | 4936947 | 150 | |
| L79948.1 | - | 4 | 363 | 150 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein sumoylation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular protein localization
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
SUMO transferase activity
ubiquitin protein ligase binding
poly(A) RNA binding
protein sumoylation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular protein localization
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
SUMO transferase activity
ubiquitin protein ligase binding
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154077 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154074 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154079 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154071 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154075 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154072 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156002 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154870 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009206 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969523 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2132865 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789504 | AK138369.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789504 | AK152200.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789504 | AK165576.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789504 | BC027536.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789504 | BC053965.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789504 | BC100462.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789504 | NM_007534.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789504 | NM_007536.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398214 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160936 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159484 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169586 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969523 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113653 | AK132213.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156002 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009206 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2132865 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154870 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427906 | AK160418.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427906 | AK167308.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427906 | BC017522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427906 | BC083326.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427906 | BC095930.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154078 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07502782 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159484 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169586 | - | - |
| Whole Mouse Genome G4122A | A_51_P427906 | AK160418.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427906 | AK167308.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427906 | BC017522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427906 | BC083326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427906 | BC095930.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124275 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136314 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169586 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159484 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09250021 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09250027 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09250026 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09250022 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154072 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154079 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154071 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154075 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154074 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28153977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09730482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09506823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09279820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09138647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09664692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09138646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154046 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10438455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353220 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484352 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10392947 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10520124 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484351 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10438454 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10353219 | - | - |
| Mouse Genome 430 2.0 Array | 1415782_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415782_at | - | - |
| Mouse Genome 430 2.0 Array | 1415781_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1415781_PM_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100038312.mm.1 | NM_133354.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038312.mm.1 | NM_133354.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038312.mm.1 | TC1100004077.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038306.mm.1 | NM_133354.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038306.mm.1 | NM_133354.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038306.mm.1 | TC1100004077.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038309.mm.1 | NM_133354.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038309.mm.1 | NM_133354.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038309.mm.1 | TC1100004077.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038304.mm.1 | NM_133354.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038304.mm.1 | NM_133354.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038304.mm.1 | TC1100004077.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038303.mm.1 | NM_133354.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038303.mm.1 | NM_133354.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038303.mm.1 | TC1100004077.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038313.mm.1 | NM_133354.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038313.mm.1 | NM_133354.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038313.mm.1 | TC1100004077.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415782_at | - | - |
| Mouse Expression Set 430 | 1415782_PM_at | - | - |
| Mouse Expression Set 430 | 1415781_PM_a_at | - | - |
| Mouse Expression Set 430 | 1415781_a_at | - | - |
| Murine Genome U74v2 Set | 96316_PM_at | - | - |
| Murine Genome U74v2 Set | 96316_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415782_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415781_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415782_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415781_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 904101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3610706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 70010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070010 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001373.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001373.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2980331 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221126 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2980331 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001373.1_2-S | - | - |

Sumo2
Genomic Map