Assay Details
| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178782.4 | NP_848897.3 | 4 - 5 | 430 | 64 | |
| GenBank mRNA | CJ176997.1 | - | 4 - 5 | 430 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcorl1
Genomic Map