Assay Details
| Gene Symbol: Parp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205239.1 | NP_001192168.1 | 19 - 20 | 2048 | 66 | |
| NM_029922.3 | NP_084198.2 | 19 - 20 | 1988 | 66 | ||
| GenBank mRNA | AK014142.1 | - | 10 - 11 | 791 | 66 | |
| BC052064.1 | - | 19 - 20 | 1986 | 66 | ||
| BC062096.1 | - | 18 - 19 | 1905 | 5716648 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp6
Genomic Map