Assay Details
| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008715.2 | NP_032741.2 | 1 - 2 | 570 | 78 | |
| GenBank mRNA | AK053385.1 | - | 1 - 2 | 594 | 78 | |
| AK149711.1 | - | 1 - 2 | 598 | 78 | ||
| AK172178.1 | - | 1 - 2 | 598 | 78 | ||
| BC059263.1 | - | 1 - 2 | 570 | 6409580 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ints6
Genomic Map