Assay Details
| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134115.2 | NP_598876.1 | 2 - 3 | 358 | 74 | |
| GenBank mRNA | AK035731.1 | - | 2 - 3 | 367 | 74 | |
| AK080179.1 | - | 2 - 3 | 367 | 74 | ||
| AK084433.1 | - | 2 - 3 | 357 | 74 | ||
| AY292399.1 | - | 2 - 3 | 222 | 74 | ||
| BC009658.1 | - | 2 - 3 | 358 | 3484764 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk38
Genomic Map