Assay Details
| Gene Symbol: Prrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102563.1 | NP_001096033.1 | 3 - 4 | 1297 | 101 | |
| GenBank mRNA | AK005319.1 | - | 2 - 3 | 286 | 101 | |
| BC053419.1 | - | 4 - 5 | 813 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010877 | BC053419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010877 | BF323287.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010877 | AK158895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010877 | NM_001102563.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010877 | BC054744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | BC053419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | AK158895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | AK005319.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | NM_001102563.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | BC054744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | BC053419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | AK158895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | AK005319.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | NM_001102563.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344461 | BC054744.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155117 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010877 | BC053419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010877 | BF323287.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010877 | AK158895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010877 | NM_001102563.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010877 | BC054744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | BC053419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | AK158895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | AK005319.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | NM_001102563.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | BC054744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | BC053419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | AK158895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | AK005319.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | NM_001102563.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344461 | BC054744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | BC053419.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | AK158895.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | AK005319.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | NM_001102563.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | BC054744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | BC053419.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | AK158895.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | AK005319.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | NM_001102563.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344461 | BC054744.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06411509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06411500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06130216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06130212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06030397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06411505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503591 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568138 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568137 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568135 | - | - |
| Mouse Genome 430 2.0 Array | 1429113_at | - | - |
| Mouse Genome 430 2.0 Array | 1429113_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700036137.mm.1 | NM_001102563.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036137.mm.1 | NM_001102563.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036137.mm.1 | TC0700004661.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036138.mm.1 | NM_001102563.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036138.mm.1 | NM_001102563.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036138.mm.1 | TC0700004661.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429113_at | - | - |
| Mouse Expression Set 430 | 1429113_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 363407 | - | - |

Prrt2
Genomic Map