Assay Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112705.2 | NP_001106176.1 | 15 - 16 | 1891 | 134 | |
| NM_001294329.1 | NP_001281258.1 | 13 - 14 | 1751 | 134 | ||
| NM_001294330.1 | NP_001281259.1 | 13 - 14 | 1699 | 134 | ||
| NM_001294331.1 | NP_001281260.1 | 15 - 16 | 1904 | 134 | ||
| NM_001294334.1 | NP_001281263.1 | 15 - 16 | 1891 | 134 | ||
| GenBank mRNA | AF045252.1 | - | 15 - 16 | 1910 | 134 | |
| AF045253.1 | - | 14 - 15 | 1338 | 134 | ||
| AF045254.1 | - | 10 - 11 | 1052 | 134 | ||
| AK014829.1 | - | 15 - 16 | 1529 | 134 | ||
| AK165667.1 | - | 15 - 16 | 1894 | 134 | ||
| AX775743.1 | - | 9 - 10 | 916 | 134 | ||
| BC066198.1 | - | 13 - 14 | 1244 | 134 | ||
| U56909.1 | - | 6 - 7 | 589 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tlk2
Genomic Map