Assay Details
| Gene Symbol: Efna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207654.2 | NP_997537.1 | 4 - 5 | 752 | 90 | |
| GenBank mRNA | BC040218.1 | - | 4 - 5 | 752 | 4954352 | 90 |
| U90664.1 | - | 4 - 5 | 579 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efna5
Genomic Map