Assay Details
| Gene Symbol: Eif4g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040131.2 | NP_001035221.1 | 4 - 5 | 581 | 133 | |
| NM_013507.3 | NP_038535.2 | 4 - 5 | 581 | 133 | ||
| GenBank mRNA | AK144309.1 | - | 4 - 5 | 555 | 133 | |
| AK150745.1 | - | 4 - 5 | 558 | 133 | ||
| AK151637.1 | - | 4 - 5 | 558 | 133 | ||
| BC039274.1 | - | 3 - 4 | 259 | 133 | ||
| BC040391.1 | - | 3 - 4 | 270 | 4018962 | 133 | |
| BC043034.1 | - | 3 - 4 | 261 | 5696921 | 133 | |
| BC056387.1 | - | 4 - 5 | 545 | 6854755 | 133 | |
| BC057673.1 | - | 5 - 6 | 433 | 4020759 | 133 | |
| BC064810.1 | - | 3 - 4 | 314 | 6825556 | 133 | |
| BC092521.1 | - | 4 - 5 | 536 | 30649919 | 133 | |
| BC127064.1 | - | 4 - 5 | 581 | 40109978 | 133 | |
| CK619644.1 | - | 2 - 3 | 171 | 133 | ||
| U63323.1 | - | 4 - 5 | 533 | 133 | ||
| U76112.1 | - | 4 - 5 | 555 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144309.1, AK150745.1, AK151637.1 transcripts.
Set Membership: |
|

Eif4g2
Genomic Map