Assay Details
| Gene Symbol: Bend5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026279.3 | NP_080555.1 | 4 - 5 | 1067 | 85 | |
| NR_033793.1 | 4 - 5 | 1149 | 85 | |||
| GenBank mRNA | AK075885.1 | - | 4 - 5 | 1067 | 85 | |
| AK156708.1 | - | 4 - 5 | 969 | 85 | ||
| AK167425.1 | - | 4 - 5 | 887 | 85 | ||
| BC037633.1 | - | 4 - 5 | 900 | 85 | ||
| BC051512.1 | - | 2 - 3 | 488 | 1383740 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bend5
Genomic Map