Assay Details
| Gene Symbol: Chic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006962.1 | - | 2 - 3 | 508 | 103 | |
| AK172289.1 | - | 2 - 3 | 501 | 103 | ||
| BC048435.1 | - | 3 - 4 | 261 | 103 | ||
| BC116971.1 | - | 2 - 3 | 318 | 40126290 | 103 | |
| BC116973.1 | - | 2 - 3 | 318 | 40126292 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chic2
Genomic Map