Assay Details
| Gene Symbol: Nufip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024205.2 | NP_001019376.1 | 2 - 3 | 2082 | 134 | |
| GenBank mRNA | AK030113.1 | - | 2 - 3 | 2084 | 134 | |
| AK122494.1 | - | 2 - 3 | 2066 | 134 | ||
| AK142954.1 | - | 2 - 3 | 2052 | 134 | ||
| AK152336.1 | - | 2 - 3 | 2053 | 134 | ||
| AK160601.1 | - | 2 - 3 | 2083 | 134 | ||
| AK170764.1 | - | 2 - 3 | 2080 | 134 | ||
More Information
Set Membership: |
|

Nufip2
Genomic Map