Assay Details
| Gene Symbol: Map2k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009157.4 | NP_033183.1 | 3 - 4 | 437 | 120 | |
| GenBank mRNA | AK036928.1 | - | 3 - 4 | 441 | 120 | |
| AK044534.1 | - | 3 - 4 | 429 | 120 | ||
| AK044698.1 | - | 3 - 4 | 429 | 120 | ||
| AK139777.1 | - | 4 - 5 | 460 | 121 | ||
| BC029833.1 | - | 1 - 2 | 188 | 3962163 | 120 | |
| U18310.1 | - | 3 - 4 | 412 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC029833.1 transcripts.
Set Membership: |
|

Map2k4
Genomic Map