Assay Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170755.2 | NP_739561.2 | 3 - 4 | 487 | 107 | |
| GenBank mRNA | AK152268.1 | - | 3 - 4 | 202 | 107 | |
| AK158940.1 | - | 3 - 4 | 257 | 107 | ||
| AK160400.1 | - | 3 - 4 | 441 | 107 | ||
| AK165957.1 | - | 3 - 4 | 466 | 107 | ||
| AK172093.1 | - | 3 - 4 | 484 | 107 | ||
| BC070406.1 | - | 3 - 4 | 479 | 5707319 | 107 | |
| BY261213.1 | - | 3 - 4 | 265 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam134a
Genomic Map