Assay Details
| Gene Symbol: Car10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028296.3 | NP_082572.1 | 2 - 3 | 1069 | 74 | |
| GenBank mRNA | AB080741.1 | - | 3 - 4 | 1025 | 74 | |
| AK012302.1 | - | 3 - 4 | 1032 | 74 | ||
| AK035758.1 | - | 2 - 3 | 304 | 74 | ||
| AK038695.1 | - | 3 - 4 | 949 | 74 | ||
| AK132222.1 | - | 2 - 3 | 910 | 74 | ||
| AK162576.1 | - | 2 - 3 | 187 | 74 | ||
| BC017606.1 | - | 2 - 3 | 261 | 4507552 | 74 | |
| BC066111.1 | - | 3 - 4 | 1025 | 30360743 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car10
Genomic Map