Assay Details
| Gene Symbol: Memo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133771.2 | NP_598532.1 | 7 - 8 | 762 | 172 | |
| GenBank mRNA | AF363447.1 | - | 8 - 9 | 824 | 172 | |
| AK159560.1 | - | 7 - 8 | 741 | 172 | ||
| BC013819.1 | - | 7 - 8 | 779 | 3489363 | 172 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Memo1
Genomic Map