Assay Details
| Gene Symbol: Angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286062.1 | NP_001272991.1 | 7 - 8 | 1711 | 75 | |
| NM_009640.4 | NP_033770.2 | 7 - 8 | 1714 | 75 | ||
| GenBank mRNA | AK088439.1 | - | 7 - 8 | 1715 | 75 | |
| AK172868.1 | - | 7 - 8 | 1531 | 75 | ||
| BC067410.1 | - | 7 - 8 | 1532 | 30436666 | 75 | |
| U83509.1 | - | 7 - 8 | 1504 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Angpt1
Genomic Map