Assay Details
| Gene Symbol: Capza2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007604.2 | NP_031630.1 | 4 - 5 | 233 | 90 | |
| GenBank mRNA | AK007550.1 | - | 4 - 5 | 232 | 90 | |
| AK008204.1 | - | 4 - 5 | 219 | 90 | ||
| AK031551.1 | - | 4 - 5 | 297 | 90 | ||
| AK150587.1 | - | 4 - 5 | 235 | 90 | ||
| AY544984.1 | - | 4 - 5 | 229 | 90 | ||
| BC082589.1 | - | 4 - 5 | 233 | 30626416 | 90 | |
| U16741.1 | - | 4 - 5 | 314 | 90 | ||
More Information
Set Membership: |
|

Capza2
Genomic Map