Assay Details
| Gene Symbol: Cald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145575.3 | NP_663550.1 | 10 - 11 | 1713 | 76 | |
| GenBank mRNA | AK148009.1 | - | 8 - 9 | 1085 | 76 | |
| BC019435.1 | - | 10 - 11 | 1672 | 5148205 | 76 | |
| CX568419.1 | - | 5 - 6 | 409 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cald1
Genomic Map