Assay Details
| Gene Symbol: Cnksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177751.2 | NP_808419.1 | 2 - 3 | 493 | 95 | |
| GenBank mRNA | AK122399.1 | - | 2 - 3 | 564 | 95 | |
| AK141544.1 | - | 2 - 3 | 564 | 95 | ||
| BC043093.1 | - | 2 - 3 | 493 | 6405389 | 95 | |
| BC060716.1 | - | 2 - 3 | 493 | 6405389 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnksr2
Genomic Map