Assay Details
| Gene Symbol: Arhgef17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081116.1 | NP_001074585.1 | 18 - 19 | 5954 | 65 | |
| GenBank mRNA | AK122250.1 | - | 18 - 19 | 2609 | 65 | |
| BC028887.1 | - | 5 - 6 | 573 | 65 | ||
| BC035332.1 | - | 9 - 10 | 1429 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef17
Genomic Map