Assay Details
| Gene Symbol: Arhgef17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081116.1 | NP_001074585.1 | 8 - 9 | 4401 | 100 | |
| GenBank mRNA | AK122250.1 | - | 8 - 9 | 1056 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef17
Genomic Map