Assay Details
| Gene Symbol: Prl7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011120.2 | NP_035250.1 | 2 - 3 | 269 | 135 | |
| GenBank mRNA | AK005421.1 | - | 2 - 3 | 266 | 135 | |
| AK005557.1 | - | 2 - 3 | 269 | 135 | ||
| AK019461.1 | - | 2 - 3 | 269 | 135 | ||
| BC099470.1 | - | 2 - 3 | 258 | 30787403 | 135 | |
| X02594.1 | - | 2 - 3 | 269 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prl7d1
Genomic Map