Assay Details
| Gene Symbol: Apc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007462.3 | NP_031488.2 | 14 - 15 | 2054 | 85 | |
| GenBank mRNA | AK046280.1 | - | 14 - 15 | 2044 | 85 | |
| AK080907.1 | - | 14 - 15 | 2054 | 85 | ||
| BC141141.1 | - | 13 - 14 | 1767 | 9055753 | 85 | |
| M88127.1 | - | 13 - 14 | 1743 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apc
Genomic Map