Assay Details
| Gene Symbol: Rnf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025999.3 | NP_080275.2 | 4 - 5 | 527 | 67 | |
| GenBank mRNA | AK006791.1 | - | 4 - 5 | 67 | ||
| AK011839.1 | - | 4 - 5 | 516 | 67 | ||
| AK015505.1 | - | 4 - 5 | 528 | 67 | ||
| AK139910.1 | - | 4 - 5 | 521 | 67 | ||
| AK154417.1 | - | 4 - 5 | 551 | 67 | ||
| BC018553.1 | - | 4 - 5 | 521 | 3582546 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf141
Genomic Map