| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800002461.mm.1 |
NM_001271544.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002461.mm.1 |
NM_001271545.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002461.mm.1 |
NM_001271546.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002461.mm.1 |
NM_001271547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002461.mm.1 |
NM_172830.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002461.mm.1 |
NM_172830.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002461.mm.1 |
TC1800000331.mm.1 |
MTA TC |