Assay Details
| Gene Symbol: Cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141737.1 | - | 4 - 5 | 639 | 67 | |
| AK168287.1 | - | 4 - 5 | 634 | 67 | ||
| AK181318.1 | - | 1 - 2 | 33 | 67 | ||
| BC004669.1 | - | 3 - 4 | 343 | 3498846 | 67 | |
| U15562.1 | - | 4 - 5 | 605 | 67 | ||
More Information
Set Membership: |
|

Cdc25c
Genomic Map