Assay Details
| Gene Symbol: Cdc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178347.4 | NP_848124.1 | 2 - 3 | 269 | 82 | |
| GenBank mRNA | AK031459.1 | - | 2 - 3 | 252 | 82 | |
| AK038523.1 | - | 2 - 3 | 252 | 82 | ||
| AK047789.1 | - | 2 - 3 | 252 | 82 | ||
| AK048246.1 | - | 2 - 3 | 252 | 82 | ||
| AK052567.1 | - | 2 - 3 | 269 | 82 | ||
| AK075948.1 | - | 2 - 3 | 270 | 82 | ||
| AK079681.1 | - | 1 - 2 | 336 | 82 | ||
| AK081461.1 | - | 2 - 3 | 270 | 82 | ||
| BC059013.1 | - | 2 - 3 | 238 | 6813730 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059013.1 transcripts.
Set Membership: |
|

Cdc23
Genomic Map