Assay Details
| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301628.1 | NP_001288557.1 | 9 - 10 | 1269 | 108 | |
| NM_027604.4 | NP_081880.2 | 10 - 11 | 1356 | 108 | ||
| GenBank mRNA | AF468037.1 | - | 10 - 11 | 1260 | 108 | |
| AK014891.1 | - | 4 - 5 | 567 | 108 | ||
| AK052398.1 | - | 8 - 9 | 1085 | 108 | ||
| AK133852.1 | - | 9 - 10 | 1178 | 108 | ||
| AK145749.1 | - | 10 - 11 | 1260 | 108 | ||
| AK161469.1 | - | 10 - 11 | 1299 | 108 | ||
| AK161957.1 | - | 9 - 10 | 1217 | 108 | ||
| AK163315.1 | - | 9 - 10 | 1198 | 108 | ||
| AK168755.1 | - | 9 - 10 | 1204 | 108 | ||
More Information
Set Membership: |
|

Usp15
Genomic Map