Assay Details
| Gene Symbol: Rsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081267.2 | NP_001074736.1 | 3 - 4 | 395 | 138 | |
| GenBank mRNA | AK032853.1 | - | 3 - 4 | 371 | 138 | |
| AK146154.1 | - | 3 - 4 | 399 | 138 | ||
| AK146675.1 | - | 3 - 4 | 405 | 138 | ||
| BC085127.1 | - | 3 - 4 | 396 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsf1
Genomic Map