Assay Details
| Gene Symbol: Dnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173383.2 | NP_775559.2 | 1 - 2 | 67 | 97 | |
| GenBank mRNA | BC034897.1 | - | 1 - 2 | 67 | 3470697 | 97 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC034897.1,NM_173383.2 transcripts.
This assay is in the 5'UTR region of the BC034897.1,NM_173383.2 transcripts.
Set Membership: |
|

Dnd1
Genomic Map