Assay Details
| Gene Symbol: Rbbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011247.2 | NP_035377.2 | 14 - 15 | 1991 | 95 | |
| GenBank mRNA | U28789.1 | - | 8 - 9 | 985 | 95 | |
| U83913.1 | - | 9 - 10 | 1134 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbbp6
Genomic Map