Assay Details
| Gene Symbol: Proc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042768.2 | NP_001036233.2 | 2 - 3 | 138 | 86 | |
| GenBank mRNA | BY788141.1 | - | 2 - 3 | 115 | 86 | |
| CA456834.1 | - | 179 | 86 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001042768.2 transcripts.
Set Membership: |
|

Proc
Genomic Map