Assay Details
| Gene Symbol: 1700074P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028550.3 | NP_082826.1 | 3 - 4 | 396 | 106 | |
| GenBank mRNA | AK006951.1 | - | 2 - 3 | 270 | 106 | |
| AK030264.1 | - | 3 - 4 | 396 | 106 | ||
| BC061035.1 | - | 2 - 3 | 307 | 6774253 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006951.1, AK030264.1, BC061035.1, NM_028550.3 transcripts.
Set Membership: |
|

1700074P13Rik
Genomic Map