Assay Details
Gene Symbol: 1700074P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028550.3 | NP_082826.1 | 4 - 5 | 571 | 71 | |
GenBank mRNA | AK006951.1 | - | 3 - 4 | 445 | 71 | |
AK030264.1 | - | 4 - 5 | 571 | 71 | ||
BC061035.1 | - | 3 - 4 | 482 | 6774253 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
