Assay Details
| Gene Symbol: Nap1l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285489.1 | NP_001272418.1 | 5 - 6 | 410 | 60 | |
| NM_001285490.1 | NP_001272419.1 | 5 - 6 | 423 | 60 | ||
| NM_008672.3 | NP_032698.1 | 5 - 6 | 410 | 60 | ||
| GenBank mRNA | AJ002198.1 | - | 5 - 6 | 380 | 60 | |
| AK090112.1 | - | 5 - 6 | 426 | 60 | ||
| AK156835.1 | - | 5 - 6 | 383 | 60 | ||
| BC089018.1 | - | 5 - 6 | 396 | 60 | ||
| BC132039.1 | - | 4 - 5 | 334 | 40130316 | 60 | |
| BC138518.1 | - | 4 - 5 | 334 | 8861538 | 60 | |
| BC145306.1 | - | 4 - 5 | 334 | 9053860 | 60 | |
| BC145308.1 | - | 4 - 5 | 334 | 9053862 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nap1l4
Genomic Map