Assay Details
| Gene Symbol: Piga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011081.2 | NP_035211.2 | 4 - 5 | 1101 | 59 | |
| GenBank mRNA | AK014488.1 | - | 2 - 3 | 2689 | 59 | |
| AK032283.1 | - | 4 - 5 | 1072 | 59 | ||
| AK161026.1 | - | 4 - 5 | 1071 | 59 | ||
| AK171453.1 | - | 4 - 5 | 694 | 59 | ||
| BC035538.1 | - | 3 - 4 | 762 | 59 | ||
| BC051166.1 | - | 3 - 4 | 322 | 59 | ||
| BC058767.1 | - | 2 - 3 | 209 | 59 | ||
| BC138755.1 | - | 3 - 4 | 1059 | 8861777 | 59 | |
| BC138758.1 | - | 3 - 4 | 1059 | 8861780 | 59 | |
| D26047.1 | - | 4 - 5 | 1070 | 59 | ||
| S78188.1 | - | 4 - 5 | 1107 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Piga
Genomic Map