Assay Details
| Gene Symbol: Cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285905.1 | NP_001272834.1 | 2 - 3 | 261 | 126 | |
| NM_001285906.1 | NP_001272835.1 | 2 - 3 | 275 | 126 | ||
| NM_001285907.1 | NP_001272836.1 | 2 - 3 | 403 | 126 | ||
| NM_027353.4 | NP_081629.1 | 2 - 3 | 255 | 126 | ||
| GenBank mRNA | AK010582.1 | - | 2 - 3 | 140 | 126 | |
| AK030393.1 | - | 2 - 3 | 137 | 126 | ||
| AK031694.1 | - | 2 - 3 | 169 | 126 | ||
| AK034689.1 | - | 2 - 3 | 179 | 126 | ||
| AK154346.1 | - | 2 - 3 | 140 | 126 | ||
| AK164102.1 | - | 1 - 2 | 119 | 126 | ||
| AK167249.1 | - | 2 - 3 | 201 | 126 | ||
| AK173122.1 | - | 1 - 2 | 97 | 126 | ||
| BC006920.1 | - | 2 - 3 | 171 | 3601764 | 126 | |
| BM944739.1 | - | 2 - 3 | 185 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd2bp2
Genomic Map