Assay Details
| Gene Symbol: Ino80e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040862.1 | - | 1 - 2 | 173 | 79 | |
| AK135531.1 | - | 1 - 2 | 140 | 79 | ||
| BC025527.1 | - | 1 - 2 | 161 | 79 | ||
| BC055928.1 | - | 2 - 3 | 407 | 3995231 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ino80e
Genomic Map