Assay Details
| Gene Symbol: Ino80e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135531.1 | - | 3 - 4 | 254 | 109 | |
| BC025527.1 | - | 3 - 4 | 275 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ino80e| Gene Symbol: Ino80e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135531.1 | - | 3 - 4 | 254 | 109 | |
| BC025527.1 | - | 3 - 4 | 275 | 109 | ||
Set Membership: |
|