Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700035999.mm.1 |
NM_153580.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035999.mm.1 |
NM_153580.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035999.mm.1 |
TC0700004301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036002.mm.1 |
NM_153580.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036002.mm.1 |
NM_153580.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036002.mm.1 |
TC0700004301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036000.mm.1 |
NM_153580.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036000.mm.1 |
NM_153580.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036000.mm.1 |
TC0700004301.mm.1 |
MTA TC |