Assay Details
| Gene Symbol: Csnk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002903.1 | - | 1 - 2 | 119 | 92 | |
| AK010730.1 | - | 1 - 2 | 116 | 92 | ||
| AK012369.1 | - | 1 - 2 | 114 | 92 | ||
| AK146186.1 | - | 1 - 2 | 139 | 92 | ||
| BC003775.1 | - | 1 - 2 | 138 | 3485097 | 92 | |
| X52959.1 | - | 1 - 2 | 119 | 92 | ||
| X56502.1 | - | 1 - 2 | 113 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002903.1,AK010730.1,AK012369.1,AK146186.1,BC003775.1,X52959.1,X56502.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Csnk2b
Genomic Map