Assay Details
| Gene Symbol: Slc44a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023557.3 | NP_076046.1 | 12 - 13 | 1144 | 88 | |
| GenBank mRNA | AK008864.1 | - | 1 - 2 | 106 | 88 | |
| AK145621.1 | - | 12 - 13 | 1136 | 88 | ||
| AK164684.1 | - | 11 - 12 | 1912 | 88 | ||
| AK166620.1 | - | 9 - 10 | 1046 | 88 | ||
| BC010808.1 | - | 12 - 13 | 1142 | 4240658 | 88 | |
| BC052652.1 | - | 12 - 13 | 1142 | 30034507 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc44a4
Genomic Map