Assay Details
| Gene Symbol: Dxo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163770.1 | NP_001157242.1 | 5 - 6 | 1148 | 71 | |
| NM_033613.2 | NP_291091.2 | 5 - 6 | 1173 | 71 | ||
| GenBank mRNA | BC004713.1 | - | 5 - 6 | 1109 | 3499698 | 71 |
| BG071924.2 | - | 2 - 3 | 71 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dxo
Genomic Map